7-Bromo-4-chlorothieno[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

070258-1G 070258-5G
101943-988EA 1687.69 USD
101943-988 101943-990
7-Bromo-4-chlorothieno[3,2-c]pyridine ≥95%
7-Bromo-4-chlorothieno[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₃BrClNS
MDL Number: MFCD11846192
CAS Number: 29064-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR