Furo[3,2-c]pyridin-4(5H)-one ≥95%

Supplier: Matrix Scientific

070257-1G 070257-5G
101943-984EA 96.92 USD
101943-984 101943-986
Furo[3,2-c]pyridin-4(5H)-one ≥95%
Furo[3,2-c]pyridin-4(5H)-one

Hazard: Irritant

Formula: C₇H₅NO₂
MW: 135.1201 g/mol
MDL Number: MFCD00183939
CAS Number: 26956-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR