5-Fluoro-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

070253-1G 070253-5G
101943-968EA 1861.54 USD
101943-968 101943-970
5-Fluoro-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
5-Fluoro-1,2,3,4-tetrahydroquinoline hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClFN
MDL Number: MFCD09026770
CAS Number: 345264-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR