tert-Butyl-2-amino-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-amino-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate

070252-1G 070252-5G
101943-964EA 1716.92 USD
101943-964 101943-966
tert-Butyl-2-amino-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate ≥95%
tert-Butyl-2-amino-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₆N₄O₂
MW: 236.28 g/mol
MDL Number: MFCD08447404
CAS Number: 1105187-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR