1H-Isoquinoline-2,4-dicarboxylic acid-2-tert-butyl ester-4-methyl ester ≥95%

Supplier: Matrix Scientific

070248-1G 070248-5G
101943-948EA 1716.92 USD
101943-948 101943-950
1H-Isoquinoline-2,4-dicarboxylic acid-2-tert-butyl ester-4-methyl ester ≥95%
1H-Isoquinoline-2,4-dicarboxylic acid-2-tert-butyl ester-4-methyl ester

Hazard: Irritant

Formula: C₁₆H₁₉NO₄
MDL Number: MFCD11973827
CAS Number: 1187830-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR