4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
070247-1G
070247-5G
101943-944EA
1687.69
USD
101943-944
101943-946
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid ≥95%
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid
Hazard: Irritant
Formula:
C₇H₃ClN₂O₂S |
MDL Number:
MFCD11846188 CAS Number: 875515-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...