2,4-Dichloro-6,7-dihydro-5H-cyclopenta[b]pyridine hydrochloride ≥95%
Supplier: Matrix Scientific
070241-1G
070241-5G
101943-920EA
1570.77
USD
101943-920
101943-922
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[b]pyridine hydrochloride ≥95%
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[b]pyridine hydrochloride
Hazard: Irritant
Formula:
C₈H₈Cl₃N |
MDL Number:
MFCD11846186 CAS Number: 1187830-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...