6-Bromopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-bromo-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one, 6-Bromopyrrolo[2,1-f][1,2,4]triazin-4(3H)-one

070239-1G 070239-5G
101943-912EA 1744.62 USD
101943-912 101943-914
6-Bromopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one ≥95%
6-Bromopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one

Hazard: Irritant

Formula: C₆H₄BrN₃O
MW: 214.03 g/mol
MDL Number: MFCD11973831
CAS Number: 888721-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR