5-Chloro-1H-imidazo[4,5-b]pyridin-2(3H)-one ≥95%

Supplier: Matrix Scientific

070237-500MG 070237-1G
101943-902EA 930.77 USD
101943-902 101943-904
5-Chloro-1H-imidazo[4,5-b]pyridin-2(3H)-one ≥95%
5-Chloro-1H-imidazo[4,5-b]pyridin-2(3H)-one

Hazard: Irritant

Formula: C₆H₄ClN₃O
MDL Number: MFCD06659688
CAS Number: 40851-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR