2-Chloro-5-iodo-1H-benzo[d]imidazole ≥95%
Supplier: Matrix Scientific
070236-1G
070236-5G
101943-898EA
1861.54
USD
101943-898
101943-900
2-Chloro-5-iodo-1H-benzo[d]imidazole ≥95%
2-Chloro-5-iodo-1H-benzo[d]imidazole
Hazard: Irritant
Formula:
C₇H₄ClIN₂ |
MDL Number:
MFCD11044655 CAS Number: 256518-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...