tert-Butyl-3-(ethoxycarbonyl)-1H-indol-2-ylcarbamate ≥95%

Supplier: Matrix Scientific

070235-1G 070235-5G
101943-894EA 1280 USD
101943-894 101943-896
tert-Butyl-3-(ethoxycarbonyl)-1H-indol-2-ylcarbamate ≥95%
tert-Butyl-3-(ethoxycarbonyl)-1H-indol-2-ylcarbamate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₄
MW: 304.35 g/mol
MDL Number: MFCD11973830
CAS Number: 1160995-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR