Ethyl-2-amino-1H-indole-3-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

070234-1G 070234-5G
101943-890EA 844.62 USD
101943-890 101943-892
Ethyl-2-amino-1H-indole-3-carboxylate hydrochloride ≥95%
Ethyl-2-amino-1H-indole-3-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
MDL Number: MFCD11973829
CAS Number: 1187830-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR