6-Aminopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one hydrochloride ≥95%

Supplier: Matrix Scientific

070232-1G 070232-5G
101943-882EA 1687.69 USD
101943-882 101943-884
6-Aminopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one hydrochloride ≥95%
6-Aminopyrrolo[1,2-f][1,2,4]triazin-4(3H)-one hydrochloride

Hazard: Irritant

Formula: C₆H₇ClN₄O
MW: 186.6 g/mol
MDL Number: MFCD28125525
CAS Number: 1160995-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR