Tetrahydro-2H-thiopyran-4-amine hydrochloride ≥95%

Supplier: Matrix Scientific

070230-1G 070230-5G
101943-874EA 815.38 USD
101943-874 101943-876
Tetrahydro-2H-thiopyran-4-amine hydrochloride ≥95%
Tetrahydro-2H-thiopyran-4-amine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNS
MDL Number: MFCD09701226
CAS Number: 233763-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR