Ethyl-2-((S)-3-amino-2-oxopiperidin-1-yl)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Ethyl 2-(3-amino-2-oxopiperidin-1-yl)acetate hydrochloride

070228-1G 070228-5G
101943-866EA 1716.92 USD
101943-866 101943-868
Ethyl-2-((S)-3-amino-2-oxopiperidin-1-yl)acetate hydrochloride ≥95%
Ethyl-2-((S)-3-amino-2-oxopiperidin-1-yl)acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O₃
MW: 236.7 g/mol
MDL Number: MFCD11973826
CAS Number: 937057-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR