Benzyl-(S)-2-oxopiperidin-3-ylcarbamate ≥95%

Supplier: Matrix Scientific

070227-1G 070227-5G
101943-862EA 1396.92 USD
101943-862 101943-864
Benzyl-(S)-2-oxopiperidin-3-ylcarbamate ≥95%
Benzyl-(S)-2-oxopiperidin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
MDL Number: MFCD14525450
CAS Number: 141136-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR