6-Bromoimidazo[1,2-a]pyridin-8-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Amino-6-bromoimidazo[1,2-a]pyridine

070223-1G 070223-5G
101943-846EA 146.15 USD
101943-846 101943-848
6-Bromoimidazo[1,2-a]pyridin-8-amine ≥95%
6-Bromoimidazo[1,2-a]pyridin-8-amine

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
MDL Number: MFCD13250043
CAS Number: 676371-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR