Ethyl-2-((tert-butoxycarbonyl)methyl)thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(((tert-butoxycarbonyl)amino)methyl)thiazole-4-carboxylate

070222-1G 070222-5G
101943-842EA 1512.31 USD
101943-842 101943-844
Ethyl-2-((tert-butoxycarbonyl)methyl)thiazole-4-carboxylate ≥95%
Ethyl-2-((tert-butoxycarbonyl)methyl)thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₄S
MW: 286.35 g/mol
MDL Number: MFCD09878704
CAS Number: 96929-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR