6,7-Dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1,5,6,7-Tetrahydropyrrolo[3,4-d]pyrimidine-2,4-dione hydrochloride

070221-1G 070221-5G
101943-838EA 1861.54 USD
101943-838 101943-840
6,7-Dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione hydrochloride ≥95%
6,7-Dihydro-1H-pyrrolo[3,4-d]pyrimidine-2,4(3H,5H)-dione hydrochloride

Hazard: Irritant

Formula: C₆H₈ClN₃O₂
MW: 189.6 g/mol
MDL Number: MFCD11616742
CAS Number: 13931-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR