2-Bromo-4-chlorothieno[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

070220-250MG 070220-500MG 070220-1G
101943-832EA 349.23 USD
101943-832 101943-834 101943-836
2-Bromo-4-chlorothieno[3,2-c]pyridine ≥95%
2-Bromo-4-chlorothieno[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₃BrClNS
CAS Number: 28948-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR