Methyl-8-aminoimidazo[1,2-a]pyridine-6-carboxylate ≥95%

Supplier: Matrix Scientific

070217-500MG 070217-1G
101943-824EA 930.77 USD
101943-824 101943-826
Methyl-8-aminoimidazo[1,2-a]pyridine-6-carboxylate ≥95%
Methyl 8-aminoimidazo[1,2-a]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₉H₉N₃O₂
MDL Number: MFCD11973821
CAS Number: 1160994-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR