tert-Butyl-4-(2-chloro-5-fluoropyrimidin-4-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

070213-1G 070213-5G
101943-808EA 1338.46 USD
101943-808 101943-810
tert-Butyl-4-(2-chloro-5-fluoropyrimidin-4-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(2-chloro-5-fluoropyrimidin-4-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉ClFN₃O₂
MDL Number: MFCD11109481
CAS Number: 1053657-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR