3-(2-Bromoimidazo[2,1-b]thiazol-6-yl)propanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

070212-1G 070212-5G
101943-804EA 1396.92 USD
101943-804 101943-806
3-(2-Bromoimidazo[2,1-b]thiazol-6-yl)propanoic acid hydrochloride ≥95%
3-(2-Bromoimidazo[2,1-b]thiazol-6-yl)propanoic acid hydrochloride

Hazard: Irritant

Formula: C₈H₈BrClN₂O₂S
MDL Number: MFCD11109476
CAS Number: 1187830-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR