tert-Butyl-3-(aminomethyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Aminomethyl)pyrrolidine, N1-BOC protected, tert-Butyl 3-(aminomethyl)pyrrolidine-1-carboxylate

070211-1G 070211-5G
101943-800EA 612.31 USD
101943-800 101943-802
tert-Butyl-3-(aminomethyl)pyrrolidine-1-carboxylate ≥95%
tert-Butyl-3-(aminomethyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD01861798
CAS Number: 270912-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR