1-tert-Butyl-4-benzyl-6-(hydroxymethyl)-1,4-diazepane-1,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

070210-500MG 070210-1G
101943-796EA 1978.46 USD
101943-796 101943-798
1-tert-Butyl-4-benzyl-6-(hydroxymethyl)-1,4-diazepane-1,4-dicarboxylate ≥95%
1-tert-Butyl-4-benzyl-6-(hydroxymethyl)-1,4-diazepane-1,4-dicarboxylate

Hazard: Irritant

Formula: C₁₉H₂₈N₂O₅
MW: 364.45 g/mol
MDL Number: MFCD11973819
CAS Number: 1105187-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR