3-Bromobenzene-1,2-diamine hydrochloride ≥95%

Supplier: Matrix Scientific

070209-1G 070209-5G
101943-792EA 815.38 USD
101943-792 101943-794
3-Bromobenzene-1,2-diamine hydrochloride ≥95%
3-Bromobenzene-1,2-diamine hydrochloride

Hazard: Irritant

Formula: C₆H₈BrClN₂
MDL Number: MFCD11518991
CAS Number: 1187830-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR