Benzyl-4-oxoazepane-1-carboxylate ≥93%
Supplier: Matrix Scientific
Synonyms:
N-Cbz-azepan-4-one, Azepan-4-one, N-CBZ protected
070208-1G
070208-5G
101943-788EA
250.77
USD
101943-788
101943-790
Benzyl-4-oxoazepane-1-carboxylate ≥93%
Benzyl-4-oxoazepane-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₇NO₃ MW: 247.2897 g/mol |
MDL Number:
MFCD03788442 CAS Number: 83621-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...