Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 7-azaindole-3-carboxylate, Methyl 1H-pyrrolo[2,3-b]pyridine-3-carboxylate

070207-500MG 070207-1G
101943-784EA 396.92 USD
101943-784 101943-786
Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylate ≥97%
Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Melting Pt: 205…206 °C
MDL Number: MFCD09864656
CAS Number: 808137-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR