Methyl-4,6-bis(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

070205-500MG 070205-1G
101943-780EA 1978.46 USD
101943-780 101943-782
Methyl-4,6-bis(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%
Methyl-4,6-bis(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₆F₆N₂O₂
MW: 312.17 g/mol
MDL Number: MFCD11982850
CAS Number: 1146081-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR