5-Chloro-6-methylpyrimidine-2,4(1H,3H)-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-6-methyluracil

070203-500MG 070203-1G
101943-772EA 960 USD
101943-772 101943-774
5-Chloro-6-methylpyrimidine-2,4(1H,3H)-dione ≥95%
5-Chloro-6-methylpyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD00466419
CAS Number: 16018-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR