5,6,7,8-Tetrahydro-4-methylpyrido[4,3-d]pyrimidine hydrochloride ≥95%

Supplier: Matrix Scientific

070200-1G 070200-5G
101943-764EA 2501.54 USD
101943-764 101943-766
5,6,7,8-Tetrahydro-4-methylpyrido[4,3-d]pyrimidine hydrochloride ≥95%
5,6,7,8-Tetrahydro-4-methylpyrido[4,3-d]pyrimidine hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MDL Number: MFCD11518994
CAS Number: 1187830-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR