5,6,7,8-Tetrahydro-4-methoxypyrido[4,3-d]pyrimidine hydrochloride ≥95%
Supplier: Matrix Scientific
070198-1G
070198-5G
101943-756EA
2093.85
USD
101943-756
101943-758
5,6,7,8-Tetrahydro-4-methoxypyrido[4,3-d]pyrimidine hydrochloride ≥95%
5,6,7,8-Tetrahydro-4-methoxypyrido[4,3-d]pyrimidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₂ClN₃O |
MDL Number:
MFCD11518993 CAS Number: 1187830-54-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...