5,6,7,8-Tetrahydro-4-methylpyrido[3,4-d]pyrimidine hydrochloride ≥95%

Supplier: Matrix Scientific

070197-1G 070197-5G
101943-752EA 2501.54 USD
101943-752 101943-754
5,6,7,8-Tetrahydro-4-methylpyrido[3,4-d]pyrimidine hydrochloride ≥95%
5,6,7,8-Tetrahydro-4-methylpyrido[3,4-d]pyrimidine hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MDL Number: MFCD11518992
CAS Number: 1187830-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR