5-Bromo-2-methylthiazole ≥97%

Supplier: Matrix Scientific

070194-1G 070194-5G 070194-25G
101943-738EA 181.54 USD
101943-738 101943-740 101943-742
5-Bromo-2-methylthiazole ≥97%
5-Bromo-2-methylthiazole

Hazard: Irritant

Formula: C₄H₄BrNS
MW: 178.06 g/mol
MDL Number: MFCD09879074
CAS Number: 57268-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR