2-Methyl-5-(tributylstannyl)pyridine ≥96%
Supplier: Matrix Scientific
070193-500MG
070193-1G
070193-5G
101943-732EA
320
USD
101943-732
101943-734
101943-736
2-Methyl-5-(tributylstannyl)pyridine ≥96%
2-Methyl-5-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NSn |
MDL Number:
MFCD12827559 CAS Number: 167556-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...