2,5-Dibromo-3-fluoropyridine ≥98%

Supplier: Matrix Scientific

070188-1G 070188-5G
101943-728EA 89.23 USD
101943-728 101943-730
2,5-Dibromo-3-fluoropyridine ≥98%
2,5-Dibromo-3-fluoropyridine

Hazard: Irritant

Formula: C₅H₂Br₂FN
MDL Number: MFCD06659497
CAS Number: 156772-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR