2-Bromo-5-chloro-3-fluoropyridine ≥97%

Supplier: Matrix Scientific

070186-1G 070186-5G
101943-718EA 73.85 USD
101943-718 101943-720
2-Bromo-5-chloro-3-fluoropyridine ≥97%
2-Bromo-5-chloro-3-fluoropyridine

Hazard: Irritant

Formula: C₅H₂BrClFN
MW: 210.44 g/mol
Boiling Pt: 77…78 °C (7 mmHg)
MDL Number: MFCD11044268
CAS Number: 514797-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR