5-Chloro-3-fluoropyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

070185-500MG 070185-1G 070185-5G
101943-710EA 815.38 USD
101943-710 101943-712 101943-714
5-Chloro-3-fluoropyridine-2-carboxylic acid ≥97%
5-Chloro-3-fluoropyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₃ClFNO₂
MDL Number: MFCD12827555
CAS Number: 207994-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR