3-Bromo-5-chloropyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-5-chloropicolinonitrile

070184-1G 070184-5G
101943-706EA 67.69 USD
101943-706 101943-708
3-Bromo-5-chloropyridine-2-carbonitrile ≥97%
3-Bromo-5-chloropyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₂BrClN₂
MW: 217.46 g/mol
MDL Number: MFCD12827549
CAS Number: 760207-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR