5-Bromo-3-chloropyridine-2-carbonitrile ≥97%
Supplier: Matrix Scientific
070183-1G
070183-5G
070183-25G
101943-700EA
60
USD
101943-700
101943-702
102551-768
5-Bromo-3-chloropyridine-2-carbonitrile ≥97%
5-Bromo-3-chloropyridine-2-carbonitrile
Hazard: Irritant
Formula:
C₆H₂BrClN₂ |
MDL Number:
MFCD12827550 CAS Number: 945557-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...