5-Bromo-4-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

070181-1G 070181-5G
101943-690EA 815.38 USD
101943-690 101943-692
5-Bromo-4-methyl-1H-indazole ≥97%
5-Bromo-4-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MDL Number: MFCD11007842
CAS Number: 1082041-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR