5-Bromo-6-methoxy-1H-indazole ≥97%

Supplier: Matrix Scientific

070180-1G 070180-5G
101943-686EA 930.77 USD
101943-686 101943-688
5-Bromo-6-methoxy-1H-indazole ≥97%
5-Bromo-6-methoxy-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂O
MDL Number: MFCD11007880
CAS Number: 152626-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR