6-Bromo-4-fluoro-1H-indazole ≥97%
Supplier: Matrix Scientific
070177-1G
070177-5G
101943-674EA
815.38
USD
101943-674
101943-676
6-Bromo-4-fluoro-1H-indazole ≥97%
6-Bromo-4-fluoro-1H-indazole
Hazard: Irritant
Formula:
C₇H₄BrFN₂ MW: 215.0200 g/mol |
MDL Number:
MFCD07781444 CAS Number: 885520-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...