5-Bromo-6-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

070176-1G 070176-5G 070176-25G
101943-668EA 52.31 USD
101943-668 101943-670 101943-672
5-Bromo-6-methyl-1H-indazole ≥97%
5-Bromo-6-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09026991
CAS Number: 885223-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR