2-Fluoro-4-methyl-5-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

070173-1G 070173-5G
101943-654EA 1106.15 USD
101943-654 101943-656
2-Fluoro-4-methyl-5-(tributylstannyl)pyridine ≥96%
2-Fluoro-4-methyl-5-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₂FNSn
MDL Number: MFCD12827558
CAS Number: 1245816-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR