5-Chloro-3-fluoropyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

070171-1G 070171-5G
101943-646EA 304.62 USD
101943-646 101943-648
5-Chloro-3-fluoropyridine-2-carbonitrile ≥97%
5-Chloro-3-fluoropyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₂ClFN₂
MDL Number: MFCD07375055
CAS Number: 207994-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR