2-Bromo-5-nitrophenol ≥98%

Supplier: Matrix Scientific

070170-1G 070170-5G
101943-640EA 63.08 USD
101943-640 101943-642
2-Bromo-5-nitrophenol ≥98%
2-Bromo-5-nitrophenol

Hazard: Irritant

Formula: C₆H₄BrNO₃
MW: 218.0100 g/mol
MDL Number: MFCD00466299
CAS Number: 52427-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR