1H-Pyrrolo[3,2-b]pyridine-4-oxide ≥97%

Supplier: Matrix Scientific

070167-500MG 070167-1G
101943-622EA 407.69 USD
101943-622 101943-624
1H-Pyrrolo[3,2-b]pyridine-4-oxide ≥97%
1H-Pyrrolo[3,2-b]pyridine-4-oxide

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD12827548
CAS Number: 1116136-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR