1-Ethoxy-1-(tributylstannyl)ethene ≥96%
Supplier: Matrix Scientific
Synonyms:
Tributyl(1-ethoxyvinyl)tin, Tri-n-butyl(1-ethoxyvinyl)tin, 1-Ethoxyvinyltri-n-butyltin
070165-1G
070165-5G
070165-25G
101943-612EA
203.08
USD
101943-612
101943-614
101943-616
1-Ethoxy-1-(tributylstannyl)ethene ≥96%
1-Ethoxy-1-(tributylstannyl)ethene
Hazard: Irritant/Refrigerate
Formula:
C₁₆H₃₄OSn Density: 1.067 Flash Pt: >110 °C |
MDL Number:
MFCD00010240 CAS Number: 97674-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...