2-Methylene-1,3-propanediol ≥96%
Supplier: Matrix Scientific
070163-1G
070163-5G
070163-25G
101943-604EA
15.38
USD
101943-604
101943-606
102551-766
2-Methylene-1,3-propanediol ≥96%
2-Methylene-1,3-propanediol
Hazard: Irritant
Formula:
C₄H₈O₂ Boiling Pt: 93…95 °C (2 mmHg) Flash Pt: 110 °C |
MDL Number:
MFCD00075162 CAS Number: 3513-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...